Learn More
Alkali Scientific ProCycle™ White Roche 480 LightCycler,qPCR Plate, Full Edge, 384 Well, 10/pk
SDP
Catalog No. 502389325
Encompass_Preferred
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-238-9325 Supplier Alkali Scientific Supplier No. PC3R
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free.