Learn More
Alkali Scientific ProCycle™ White Roche 480 LightCycler, qPCR Plate, Full Edge, 96 Well, 10/pk
SDP
Catalog No. 502389009
Encompass_Preferred
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Crystal-clear wells to easily verify sample volume. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-238-9009 Supplier Alkali Scientific Supplier No. PC9R
Broad thermal cycler compatibility. High-contrast, black alphanumeric indexing for easy well identification. Crystal-clear wells to easily verify sample volume. Ultra-thin, uniform wells ensure optimal heat transfer for high reaction efficiency. Well ridges reduce sample evaporation when sealing with film, foil, or strip caps. Manufactured from high-clarity, ultra-pure polypropylene (meets FDA 21 CFR 177.1520 & USP Class VI). For use in pharmaceutical industry, food industry, and molecular biology. Certified free of detectable DNase, RNase, DNA, PCR inhibitors, and tested pyrogen-free.