Learn More
AdipoGen Procyclidine hydrochloride
SDP
Catalog No. 502560172
Encompass_Preferred
Chemical. CAS 1508-76-5. Formula C19H29NO . HCl. MW 323.9. Procyclidine is a synthetic anticholinergic agent and atropine-like antispasmodic. It exhibits therapeutic effects against parkinsons disease and schizophrenia. Procyclidine is an antagonist of muscarinic acetylcholine receptors Kis = 4.47, 42.66, 7.24, 6.03, and 4.79 nM for M1-5 receptors, respectively. It also affects N-methyl-D-aspartate NMDA glutamine and nicotinic acetylcholine receptors to a lower extent.
Catalog No. 50-256-0172 Supplier AdipoGen Supplier No. CDXP0608G005
Chemical. CAS 1508-76-5. Formula C19H29NO . HCl. MW 323.9. Procyclidine is a synthetic anticholinergic agent and atropine-like antispasmodic. It exhibits therapeutic effects against parkinsons disease and schizophrenia. Procyclidine is an antagonist of muscarinic acetylcholine receptors Kis = 4.47, 42.66, 7.24, 6.03, and 4.79 nM for M1-5 receptors, respectively. It also affects N-methyl-D-aspartate NMDA glutamine and nicotinic acetylcholine receptors to a lower extent.