Learn More
Medchemexpress LLC Procymidone | 32809-16-8 | 99.64% | 250 MG
SDP
Catalog No. 5000414114
Encompass_Preferred
Procymidone is a pesticide and an immunogen hapten that can be combined with cOVA (carrier protein) to produce polyclonal antibodies. It is recognized for its antifungal activity, targeting fungal pathways, making it useful in anti-infection research.
  • Can be combined with cOVA to produce polyclonal antibodies.
  • Exhibits antifungal activity.
  • Soluble in DMSO at 23.33 mg/mL.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-14114 Supplier Medchemexpress LLC Supplier No. 250MGHYB1884
Add to Cart
Add to Cart
Procymidone is a pesticide and an immunogen hapten that can be combined with cOVA (carrier protein) to produce polyclonal antibodies. It is recognized for its antifungal activity, targeting fungal pathways, making it useful in anti-infection research.
  • Can be combined with cOVA to produce polyclonal antibodies.
  • Exhibits antifungal activity.
  • Soluble in DMSO at 23.33 mg/mL.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.