Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Procymidox-d5 | 00-00-0 | C13H6D5Cl2NO2 | 10MG
SDP

Catalog No. 5000261017
Encompass_Preferred

Procymidox-d5 is the deuterium-labeled analogue of Procymidox supplied for analytical and tracer applications. It is intended for use as an internal standard in quantitative workflows; batch-specific purity and storage instructions are provided in the certificate of analysis.

  • Isotope-labeled internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Useful as a tracer in analytical experiments
  • Molecular formula C13H6D5Cl2NO2; molecular weight 289.17
  • Purity and storage conditions specified on the certificate of analysis

Catalog No. 50-002-61017 Supplier Medchemexpress LLC Supplier No. HY142813S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procymidox-d5 is the deuterium-labeled analogue of Procymidox supplied for analytical and tracer applications. It is intended for use as an internal standard in quantitative workflows; batch-specific purity and storage instructions are provided in the certificate of analysis.

  • Isotope-labeled internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Useful as a tracer in analytical experiments
  • Molecular formula C13H6D5Cl2NO2; molecular weight 289.17
  • Purity and storage conditions specified on the certificate of analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.