Learn More
Medchemexpress LLC Procymidox-d5 | 32809-16-8 | C13H6D5Cl2NO2 | 1 MG
SDP
Catalog No. 5000353598
Encompass_Preferred
Procymidox-d5 is the deuterium labeled Procymidox. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-53598 Supplier Medchemexpress LLC Supplier No. HY142813S1MG
Procymidox-d5 is the deuterium labeled Procymidox. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS