Learn More
Medchemexpress LLC Procymidox-d5 | >95.0% | 289.17 | C13H6D5Cl2NO2 | 5 MG

Supplier: Medchemexpress LLC HY142813S5MG
Procymidox-d5 is the deuterium-labeled analogue of procymidone supplied as an isotope-labeled reference for analytical workflows. It is intended for use as a tracer or internal standard to improve accuracy and precision in quantitative NMR, GC-MS, and LC-MS measurements, compensating for extraction and ionization variability in sample analysis.
- Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS quantitation.
- Isotope composition reflected in molecular formula C13H6D5Cl2NO2.
- Molecular weight 289.17.
- Purity >95.0% (HPLC) as stated by supplier listings.
- Supplied as a small research-grade vial (5 MG) for analytical use.
- Store under conditions specified on the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.