Learn More
Cayman Chemical Procyanidin A1, Purity: ≥98%, CAS: 103883-03-0, 10 mg
SDP
Catalog No. 502918884
Encompass_Preferred
A polyphenol flavonoid dimer with diverse biological activities; binds to PlGF and VEGF (Kds = 394 and 476 nM, respectively); increases the activity of JAK2 (EC50 = 660 nM); scavenges superoxide radicals (IC50 = 14.7 µM); inhibits the cytopathic effects of HIV-1 in MT-4 cells (IC50 = 14 µg/ml); increases the proliferation of isolated mouse splenocytes and peritoneal macrophages from 25-100 µM and DAMI megakaryoblasts (EC50 = 8.58 µM); prevents decreases in the number of platelets in peripheral blood and megakaryocytes in the bone marrow in a mouse model of carboplatin-induced thrombocytopenia at 50 mg/kg
Catalog No. 50-291-8884 Supplier Cayman Chemical Supplier No. 3889310 MG
A polyphenol flavonoid dimer with diverse biological activities; binds to PlGF and VEGF (Kds = 394 and 476 nM, respectively); increases the activity of JAK2 (EC50 = 660 nM); scavenges superoxide radicals (IC50 = 14.7 µM); inhibits the cytopathic effects of HIV-1 in MT-4 cells (IC50 = 14 µg/ml); increases the proliferation of isolated mouse splenocytes and peritoneal macrophages from 25-100 µM and DAMI megakaryoblasts (EC50 = 8.58 µM); prevents decreases in the number of platelets in peripheral blood and megakaryocytes in the bone marrow in a mouse model of carboplatin-induced thrombocytopenia at 50 mg/kg