Learn More
Cayman Chemical Procyanidin C1, Purity: ≥98%, CAS: 37064-30-5, 10 mg
SDP
Catalog No. 502934448
Encompass_Preferred
A polyphenol flavonoid with HIV-related and antioxidant activities; activates latent HIV-1 provirus in JLR2 cells at 12.5-50 µM; scavenges DPPH radicals (IC50 = 3.2 µg/ml); inhibits α-glucosidase activity (IC50 = 3.8 µg/ml); inhibits 15-LO activity (IC50 = 57.6 µg/ml); prevents phosphorylation of ERK1/2 and the production of ROS in THP-1 cells at 10 µg/ml
Catalog No. 50-293-4448 Supplier Cayman Chemical Supplier No. 2520110 MG
A polyphenol flavonoid with HIV-related and antioxidant activities; activates latent HIV-1 provirus in JLR2 cells at 12.5-50 µM; scavenges DPPH radicals (IC50 = 3.2 µg/ml); inhibits α-glucosidase activity (IC50 = 3.8 µg/ml); inhibits 15-LO activity (IC50 = 57.6 µg/ml); prevents phosphorylation of ERK1/2 and the production of ROS in THP-1 cells at 10 µg/ml