Learn More
Phenomenex Inc Phenomenex LC Column, ODS-3, 5 μm, 150 x 4.6 mm
Catalog No. NC3713218 Shop All Phenomenex Products
Encompass
A reversed-phase HPLC column using an ODS-3 stationary phase with 5 μm particles and a 100 Å pore size, supplied in a 150 x 4.6 mm analytical format. Designed for routine analytical separations, the column delivers robust retention and reproducible performance for small molecules, method development, and quality control applications.
  • 5 μm particle size for a balance of efficiency and sample capacity.
  • 100 Å pore size optimized for small molecule separations.
  • ODS-3 (octadecylsilane) stationary phase for strong reversed-phase retention.
  • 150 x 4.6 mm format compatible with standard analytical HPLC systems.
  • Suitable for routine method development, quality control, and analytical assays.
  • Confirm pH range and solvent compatibility from the manufacturer datasheet.
Catalog No. NC3713218 Supplier Phenomenex Inc Supplier No. 00F4097EO
A reversed-phase HPLC column using an ODS-3 stationary phase with 5 μm particles and a 100 Å pore size, supplied in a 150 x 4.6 mm analytical format. Designed for routine analytical separations, the column delivers robust retention and reproducible performance for small molecules, method development, and quality control applications.
  • 5 μm particle size for a balance of efficiency and sample capacity.
  • 100 Å pore size optimized for small molecule separations.
  • ODS-3 (octadecylsilane) stationary phase for strong reversed-phase retention.
  • 150 x 4.6 mm format compatible with standard analytical HPLC systems.
  • Suitable for routine method development, quality control, and analytical assays.
  • Confirm pH range and solvent compatibility from the manufacturer datasheet.