Learn More
Thermo Scientific Dionex™ Retention Time Standards for Ion Chromatography
Catalog No. NC1734188 Shop All Thermo Scientific Products
Encompass
For Use With Routine Anion Determinations | Type: Standard | Standard Components Fluoride 2 mg/L, Chloride 24 mg/L, Nitrite 16 mg/L, Bromide 36 mg/L, Nitrate36 mg/L, Phosphate 26 mg/L, Sulfate 40 mg/L | Volume (Metric) 50 mL
This item is not returnable. View return policy
Catalog No. NC1734188 Supplier Thermo Scientific Supplier No. 088957
For Use With Routine Anion Determinations | Type: Standard | Standard Components Fluoride 2 mg/L, Chloride 24 mg/L, Nitrite 16 mg/L, Bromide 36 mg/L, Nitrate36 mg/L, Phosphate 26 mg/L, Sulfate 40 mg/L | Volume (Metric) 50 mL