Learn More
Thermo Scientific Dionex IC PEEK Viper Fitting | 0.18mm/0.007"""" x 165mm
Catalog No. NC1823761 Shop All Thermo Scientific Products
Encompass
Thermo Scientific Dionex IC PEEK Viper Fittings provide “worry-free”, finger-tight connections with virtually zero dead volume. The Dionex IC PEEK Viper fitting system ensures a correct tubing face seal, ensuring consistently low volume connections without the use of tools.They are compatible with all current Thermo Scientific Dionex IC systems, columns, and consumables.
Catalog No. NC1823761 Supplier Thermo Scientific Supplier No. 088807
Thermo Scientific Dionex IC PEEK Viper Fittings provide “worry-free”, finger-tight connections with virtually zero dead volume. The Dionex IC PEEK Viper fitting system ensures a correct tubing face seal, ensuring consistently low volume connections without the use of tools.They are compatible with all current Thermo Scientific Dionex IC systems, columns, and consumables.