Learn More
AdipoGen Prodigiosin (1 mg)
SDP
Supplier:  AdipoGen AGCN20105M001
Encompass_Preferred
Prodigiosin (1 mg), Adipogen Life Sciences. Chemical. CAS: 82-89-3. Formula: C20H25N3O. MW: 323.4. Isolated from gamma-proteobacterium strain MS-02-063. Red-pigmented antibiotic. Antimalarial. Immunosuppressive. Apoptosis inducer. Cytotoxic. Induces phosphorylation of p38-MAPK. Antimetastatic. Tumor invasion inhibitor. Induces p21WAF1/CIP1 expression. Anticancer compound. gp91(phox) and iNOS expression inhibitor. mTORC1 and mTORC2 complex inhibitor. Down-regulates SKP2 to induce p27(KIP1) stabilization.
Catalog No. 50-168-7277