Learn More
Enzo Life Sciences Prodigiosin (1mg). CAS: 82-89-3
SDP
Supplier:  Enzo Life Sciences ALX380321M001
Encompass_Preferred
Antibacterial, antifungal, cytotoxic and antialgal activity. Has been shown to inhibit NADPH oxidase activation in macrophages. Induces apoptosis by phosphorylation of p38 MAP kinase. Additionally, inhibits the association of p47-phox and Rac (NADPH oxidase cytosolic components) to the plasma membrane in macrophages. Alternative name: Prodigiosine, 4-Methoxy-5-[(5-methyl-4-pentyl-2H-pyrrol-2-ylidene)methyl]-2,2'-bi[1H-pyrrole]. Purity: ≥95% (HPLC). Appearance: Yellow to dark red solid. Solubility: Soluble in dioxane, pyridine, methylene chloride, hexane, chloroform or methanol; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9031