Learn More
AdipoGen Prodigiosin
SDP
Supplier:  AdipoGen AGCN20105C250
Encompass_Preferred
Chemical. CAS 82-89-3. Formula C20H25N3O. MW 323.4. Isolated from gamma-proteobacterium strain MS-02-063. Red-pigmented antibiotic. Antimalarial. Immunosuppressive. Apoptosis inducer. Cytotoxic. Induces phosphorylation of p38-MAPK. Antimetastatic. Tumor invasion inhibitor. Induces p21WAF1/CIP1 expression. Anticancer compound. gp91phox and iNOS expression inhibitor. mTORC1 and mTORC2 complex inhibitor. Down-regulates SKP2 to induce p27KIP1 stabilization.
Catalog No. 50-256-2657