Learn More
Abcam Prodigiosin, apoptotic agent, 1MG
Catalog No. 502705874
Encompass_Preferred
MW 323.4 Da, Purity >95%. Potent apoptotic agent. Antibiotic agent. Induces caspase-dependent apoptosis. Inhibits mTORC1 and mTORC2. Reactivates p53. Inhibits iNOS expression. Shows anti-ischemic and antimalarial effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5874 Supplier Abcam Supplier No. AB1443311MG
MW 323.4 Da, Purity >95%. Potent apoptotic agent. Antibiotic agent. Induces caspase-dependent apoptosis. Inhibits mTORC1 and mTORC2. Reactivates p53. Inhibits iNOS expression. Shows anti-ischemic and antimalarial effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement