Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prodigiosin hydrochloride | 56144-17-3 | 98.1% | 359.89 g/mol | C20H26ClN3O | 1 MG
SDP

Catalog No. 5000214476
Encompass_Preferred

Prodigiosin hydrochloride is the hydrochloride salt of prodigiosin, a red-pigmented secondary metabolite used as a research reagent. It exhibits proapoptotic activity and inhibits Wnt/β-catenin signaling, with reported antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer effects.

  • High purity: 98.05% (manufacturer reported).
  • Molecular formula: C20H26ClN3O; molecular weight: 359.89 g/mol.
  • Appearance: solid, purple to purplish red.
  • Supplied in small research quantities suitable for in vitro studies.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Common applications: apoptosis studies and modulation of Wnt/β-catenin signaling.

Catalog No. 50-002-14476 Supplier Medchemexpress LLC Supplier No. HY100711A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prodigiosin hydrochloride is the hydrochloride salt of prodigiosin, a red-pigmented secondary metabolite used as a research reagent. It exhibits proapoptotic activity and inhibits Wnt/β-catenin signaling, with reported antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer effects.

  • High purity: 98.05% (manufacturer reported).
  • Molecular formula: C20H26ClN3O; molecular weight: 359.89 g/mol.
  • Appearance: solid, purple to purplish red.
  • Supplied in small research quantities suitable for in vitro studies.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
  • Common applications: apoptosis studies and modulation of Wnt/β-catenin signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.