Learn More
Medchemexpress LLC Prodigiosin hydrochloride | 56144-17-3 | 98.1% | 359.89 g/mol | C20H26ClN3O | 100 UG
SDP
Catalog No. 5000213846
Encompass_Preferred
Prodigiosin hydrochloride is a research-use small-molecule hydrochloride salt reported to have antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer activities. It is supplied as a solid, purple to purplish red material intended for in vitro studies; follow appropriate handling and storage guidelines.
Catalog No. 50-002-13846 Supplier Medchemexpress LLC Supplier No. HY100711A100UG
Prodigiosin hydrochloride is a research-use small-molecule hydrochloride salt reported to have antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer activities. It is supplied as a solid, purple to purplish red material intended for in vitro studies; follow appropriate handling and storage guidelines.