Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prodigiosin hydrochloride | 56144-17-3 | 98.1% | 359.89 g/mol | C20H26ClN3O | 250 UG
SDP

Catalog No. 5000214330
Encompass_Preferred

Prodigiosin hydrochloride is the hydrochloride salt of the natural product prodigiosin, provided as a research reagent with reported antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer activities. It is supplied as a solid for use in biochemical and cell biology studies and analytical assays.

  • Small-molecule hydrochloride salt with reported broad-spectrum bioactivity.
  • High purity suitable for research use (≈98.1%).
  • Solid, purple to purplish red appearance.
  • Molecular weight 359.89 g/mol.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • Available in microgram and milligram quantities for assay development.

Catalog No. 50-002-14330 Supplier Medchemexpress LLC Supplier No. HY100711A250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prodigiosin hydrochloride is the hydrochloride salt of the natural product prodigiosin, provided as a research reagent with reported antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer activities. It is supplied as a solid for use in biochemical and cell biology studies and analytical assays.

  • Small-molecule hydrochloride salt with reported broad-spectrum bioactivity.
  • High purity suitable for research use (≈98.1%).
  • Solid, purple to purplish red appearance.
  • Molecular weight 359.89 g/mol.
  • Store sealed at -20°C; in solution store at -80°C for long-term stability.
  • Available in microgram and milligram quantities for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.