Learn More
Medchemexpress LLC Prodigiosin hydrochloride | 56144-17-3 | 98.1% | 359.89 g/mol | C20H26ClN3O | 500 UG
SDP
Catalog No. 5000214329
Encompass_Preferred
Prodigiosin hydrochloride is the hydrochloride salt of prodigiosin, a bioactive natural product reported to have antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer effects. It is supplied for research use in cell biology and biochemical assays investigating apoptosis and signaling pathways.
  • Bioactive small molecule with multiple reported biological activities.
  • Useful for cell biology and biochemical assays, including apoptosis studies.
  • High purity (98.1%) for reproducible experimental results.
  • Available in small research pack sizes such as 500 μg for screening and optimization.
  • Storage recommendations: solid at -20°C; in solvent at -80°C for long-term stability.
Catalog No. 50-002-14329 Supplier Medchemexpress LLC Supplier No. HY100711A500UG
Prodigiosin hydrochloride is the hydrochloride salt of prodigiosin, a bioactive natural product reported to have antibacterial, antifungal, antiprotozoal, antimalarial, immunosuppressive, and anticancer effects. It is supplied for research use in cell biology and biochemical assays investigating apoptosis and signaling pathways.
  • Bioactive small molecule with multiple reported biological activities.
  • Useful for cell biology and biochemical assays, including apoptosis studies.
  • High purity (98.1%) for reproducible experimental results.
  • Available in small research pack sizes such as 500 μg for screening and optimization.
  • Storage recommendations: solid at -20°C; in solvent at -80°C for long-term stability.