Learn More
Medchemexpress LLC HY-101605 1mg Medchemexpress, Prodipine hydrochloride CAS:31314-39-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016051MG
Encompass_Preferred
Medchemexpress, HY-101605 1mg Prodipine HCl CAS:31314-39-3 Prodipine, a diphenyl-phosphonate derivative. The IC50s of Prodipine for purified and plasma Dipeptidyl peptidase IV (DPP IV) from the rabbit are 4.5 μM and 30 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7861