Learn More
Medchemexpress LLC Prodipine hydrochloride | 31314-39-3 | 99.8% | 315.88 g/mol | C20H26ClN | 5 MG
SDP
Catalog No. 5000248959
Encompass_Preferred
Prodipine hydrochloride is a diphenyl-phosphonate derivative that acts as a dipeptidyl peptidase IV (DPP-IV) inhibitor. Supplied as the hydrochloride salt for research use, it is intended for enzymology, inhibitor screening, and preclinical biochemical assays; reported IC50 values are ~4.5 μM (purified rabbit DPP-IV) and ~30 μM (rabbit plasma).
  • Potent DPP-IV inhibitory activity in vitro.
  • Supplied as hydrochloride salt for improved solubility.
  • High purity suitable for biochemical assays.
  • Available in small milligram pack sizes for screening.
  • Characterized with reported IC50 data for assay comparison.
Catalog No. 50-002-48959 Supplier Medchemexpress LLC Supplier No. HY1016055MG
Prodipine hydrochloride is a diphenyl-phosphonate derivative that acts as a dipeptidyl peptidase IV (DPP-IV) inhibitor. Supplied as the hydrochloride salt for research use, it is intended for enzymology, inhibitor screening, and preclinical biochemical assays; reported IC50 values are ~4.5 μM (purified rabbit DPP-IV) and ~30 μM (rabbit plasma).
  • Potent DPP-IV inhibitory activity in vitro.
  • Supplied as hydrochloride salt for improved solubility.
  • High purity suitable for biochemical assays.
  • Available in small milligram pack sizes for screening.
  • Characterized with reported IC50 data for assay comparison.