Learn More
Medchemexpress LLC Prodipine hydrochloride | 31314-39-3 | 99.8% | 315.9 g/mol | C20H26ClN | 10 MG
SDP
Catalog No. 5000248962
Encompass_Preferred
Prodipine hydrochloride is a diphenyl-phosphonate derivative used as a research DPP-IV inhibitor. It has been characterized in vitro and in vivo for inhibition of dipeptidyl peptidase IV and is supplied for laboratory research use.
  • Inhibits dipeptidyl peptidase IV (DPP-IV) in vitro and in vivo.
  • Reported IC50 values: purified rabbit DPP-IV 4.5 μM; rabbit plasma DPP-IV 30 μM.
  • High reported purity (99.8%).
  • Molecular formula C20H26ClN; molecular weight 315.9 g/mol.
  • Intended for biochemical and pharmacological research use only.
  • Available in small research pack sizes suitable for assay work.
Catalog No. 50-002-48962 Supplier Medchemexpress LLC Supplier No. HY10160510MG
Prodipine hydrochloride is a diphenyl-phosphonate derivative used as a research DPP-IV inhibitor. It has been characterized in vitro and in vivo for inhibition of dipeptidyl peptidase IV and is supplied for laboratory research use.
  • Inhibits dipeptidyl peptidase IV (DPP-IV) in vitro and in vivo.
  • Reported IC50 values: purified rabbit DPP-IV 4.5 μM; rabbit plasma DPP-IV 30 μM.
  • High reported purity (99.8%).
  • Molecular formula C20H26ClN; molecular weight 315.9 g/mol.
  • Intended for biochemical and pharmacological research use only.
  • Available in small research pack sizes suitable for assay work.