Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human EGF Protein

Catalog No. p-7083350
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD23610 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRD23610 Supplier R&D Systems™ Supplier No. PRD23610

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Supplied as ready-to-use 5 ug protein balls

Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415.

The ED50 for this effect is 20-100 pg/mL.

Specifications

Accession Number P01133
For Use With (Application) Bioactivity
Gene ID (Entrez) 1950
Molecular Weight (g/mol) M.W. (Observed): 6 kDa, reducing conditions; M.W. (theoretical): 6 kDa
Quantity 50 μg
Source E. coli-derived human EGF protein Asn971 - Arg1023, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2°C to 8°C as supplied. 1 month at 2°C to 8°C under sterile conditions after reconstitution. 3 months (at -20°C to -80°C under sterile conditions after reconstitution.)
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG, Urogastrone
Common Name EGF
Gene Symbol EGF
Biological Activity Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED50 for this effect is typically 20-100 pg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 500 μg/mL in sterile PBS, or simply roll ProDot™ directly into cell culture medium for immediate use.
Expression System E. coli-derived Asn971 - Arg1023, with an N-terminal Met
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein EGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.