Learn More
Bachem (Pyr1)-Apelin-13 (human, bovine, mouse, rat) Acetate
SDP
Supplier:  Bachem 4104812.0005
Encompass_Preferred
(Pyr1)-Apelin-13 (human, bovine, mouse, rat) Acetate, Pyr-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH, 217082-60-5, C69H108N22O16S, Mr 1533.82, 5mg. (Glp1)-Apelin-13, human, bovine.
Catalog No. 50-194-6906