Learn More
Electron Microscopy Sciences Style 2AB, Material: PVDF., 1/EA
SDP
Catalog No. 502091055
Encompass_Preferred
The handles are made from Antimagnetic Steel and tips are synthetic fiber. Temperature Resistance: 375 degree F (190 degree C). Material: PVDF.
Catalog No. 50-209-1055 Supplier Electron Microscopy Sciences Supplier No. 78085P
The handles are made from Antimagnetic Steel and tips are synthetic fiber. Temperature Resistance: 375 degree F (190 degree C). Material: PVDF.