Learn More
Electron Microscopy Sciences M5E.CX, OAL: 80 mm. Tips: Straight, extra fine, superior finish., 1/EA
SDP
Catalog No. 502091097
Encompass_Preferred
These mini biology tweezers are the perfect choice for use under a microscope. Made of superalloy (CX), they resist extreme temperatures, chemicals and other harsh conditions and the ultra-fine tips are ideal for handling extremely minute material and grids. Anti-acid and Anti-magnetic. OAL: 80 mm. Tips: Straight, extra fine, superior finish .
Catalog No. 50-209-1097 Supplier Electron Microscopy Sciences Supplier No. 78325M5E
These mini biology tweezers are the perfect choice for use under a microscope. Made of superalloy (CX), they resist extreme temperatures, chemicals and other harsh conditions and the ultra-fine tips are ideal for handling extremely minute material and grids. Anti-acid and Anti-magnetic. OAL: 80 mm. Tips: Straight, extra fine, superior finish .