Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100L with 4 Top Drains, 3D Single-Use BPC
GREENER_CHOICE

Catalog No. SH3065302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3065302 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3065302 Supplier Thermo Scientific™ Supplier No. SH30653.02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports and 1 bottom port. The 3D square tube design with top dispense is for use with flat-bottom Thermo Scientific™ drums.

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 2 top ports and 1 bottom port. The 3D square tube design with top dispense is for use with flat-bottom Thermo Scientific™ drums.

Features:

  • Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
  • All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
  • All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:

  • Hydration and sterilization of process buffers, liquids, and culture media.
  • Chromatography feed and fraction collection.
  • Storage and transport of bulk drug product and bulk drug precursors.
  • Harvest from and feed into bioreactors and fermentors.
  • Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry's largest component library – with over 20,000 unique components that include:

  • Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
  • Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
  • Filters—used for bioburden reduction; sterilization grade filters and vent filters.
  • Tubing—thermoplastic elastomers, platinum-cured silicone.
  • Sample ports—septum-style and needle-free ports.
  • Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 L
Film Description CX5-14
Port Detail Top Ports: 2 x 0.25 in. 2 x 0.5 in.
For Use With (Equipment) SV50076.03
Outer Packaging Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity 100 L
Sterilization Method Gamma Irradiated
Tubing Line 1 MPX insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 x 0.75 in.) with dip tube length dependent on size
Tubing Line 2 Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 3 Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 4 MPX body, polycarbonate Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 12.7 x 19.1 mm (0.5 in. x 0.75 in.) dip tube length: 30.5 cm (12 in.)
Product Line Productainer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.