Learn More
Expression Systems Production Boost Additive, 100ml
SDP
Catalog No. NC0137111
Encompass_Preferred
Expression Systems Production Boost Additive (PBA) is a nutrient boost designed to increase protein expression by supporting the increased needs of insect cells infected by recombinant baculovirus. PBA is a serum-free, animal-free formulation that complements ESF 921 and ESF AF insect cell culture media and extends the late and very-late expression period of production by maintaining high culture viability. PBA is a cocktail of nutrients designed for supplementation at a rate of1–10% at 6–36 hours post infection.
• Serum-free, animal-free formulation works seamlessly with ESF 921 and ESF AF
• Extends culture period by maintaining cell viability
• Increases protein expression
• Available in 100 mL or 1 L bottles
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC0137111 Supplier Expression Systems Supplier No. 95006100
Expression Systems Production Boost Additive (PBA) is a nutrient boost designed to increase protein expression by supporting the increased needs of insect cells infected by recombinant baculovirus. PBA is a serum-free, animal-free formulation that complements ESF 921 and ESF AF insect cell culture media and extends the late and very-late expression period of production by maintaining high culture viability. PBA is a cocktail of nutrients designed for supplementation at a rate of1–10% at 6–36 hours post infection.
• Serum-free, animal-free formulation works seamlessly with ESF 921 and ESF AF
• Extends culture period by maintaining cell viability
• Increases protein expression
• Available in 100 mL or 1 L bottles
To download Instructions for Use and/or COA please visit: www.expressionsystems.com