Learn More
Med Vet International Prody Drainage/Catheter Holder, 2", J1329, Each
Catalog No. 5000581118
Encompass_Preferred
Designed to hold suction reservoir drains, other drains, and enteral feeding lines in place. - Can wrap around chest and abdomen - One size fits all, trim as necessary - Patient can move freely and comfortably - Can hold up to four drains - Blue Velcro strap locks onto drain attachment loop - Hand wash with warm water and detergent after use the air dry - U.S. Patent 6,610,032 2" wide strap
Catalog No. 50-005-81118 Supplier Med Vet International Supplier No. JOR1329
Designed to hold suction reservoir drains, other drains, and enteral feeding lines in place. - Can wrap around chest and abdomen - One size fits all, trim as necessary - Patient can move freely and comfortably - Can hold up to four drains - Blue Velcro strap locks onto drain attachment loop - Hand wash with warm water and detergent after use the air dry - U.S. Patent 6,610,032 2" wide strap