Learn More
Medchemexpress LLC Profenofos | 41198-08-7 | 96.1% | 250 MG
SDP
Catalog No. 5000406813
Encompass_Preferred
Profenofos is an insecticide utilized on field, vegetable, and fruit crops. It functions as an acetylcholinesterase (AChE) inhibitor and possesses neurotoxicity. It is for research use only.
  • Utilized on field, vegetable, and fruit crops.
  • Functions as an acetylcholinesterase (AChE) inhibitor.
  • Possesses neurotoxicity.
  • Exhibits cytotoxicity in human peripheral blood lymphocytes.
  • Induces chromosome aberrations.
  • Causes single-stranded DNA breakage.
  • Causes toxicity in mouse liver, spleen, brain, and kidney.
Catalog No. 50-004-06813 Supplier Medchemexpress LLC Supplier No. 250MGHYB0832
Profenofos is an insecticide utilized on field, vegetable, and fruit crops. It functions as an acetylcholinesterase (AChE) inhibitor and possesses neurotoxicity. It is for research use only.
  • Utilized on field, vegetable, and fruit crops.
  • Functions as an acetylcholinesterase (AChE) inhibitor.
  • Possesses neurotoxicity.
  • Exhibits cytotoxicity in human peripheral blood lymphocytes.
  • Induces chromosome aberrations.
  • Causes single-stranded DNA breakage.
  • Causes toxicity in mouse liver, spleen, brain, and kidney.