Learn More
MSC Motorola Solutions Two-way Radio, Analog UHF, 4 Channel, 2 W
Catalog No. NC2043851
Encompass
The RMU2040 two-way business radio is designed for instant communication in high-noise and harsh environments. It offers clear audio and robust durability for demanding conditions.
  • Operates on 99 UHF business exclusive frequencies with 219 PL/DPL codes.
  • Features a powerful 1500 mW speaker for clear communication.
  • Rugged design meets military specifications for sealing against dust, wind, shock, and vibration.
  • Channel announcement with voice alias for channel identification without display.
  • Provides 2 watt UHF radio coverage up to 250,000 sq. ft. or 20 floors.
  • Advanced Voice Activation (VOX) enables hands-free operation.
  • Antimicrobial coating helps prevent the growth of mold and germs.
  • Equipped with a tricolor LED interface for status identification.
Catalog No. NC2043851 Supplier MSC Supplier No. 51295467
The RMU2040 two-way business radio is designed for instant communication in high-noise and harsh environments. It offers clear audio and robust durability for demanding conditions.
  • Operates on 99 UHF business exclusive frequencies with 219 PL/DPL codes.
  • Features a powerful 1500 mW speaker for clear communication.
  • Rugged design meets military specifications for sealing against dust, wind, shock, and vibration.
  • Channel announcement with voice alias for channel identification without display.
  • Provides 2 watt UHF radio coverage up to 250,000 sq. ft. or 20 floors.
  • Advanced Voice Activation (VOX) enables hands-free operation.
  • Antimicrobial coating helps prevent the growth of mold and germs.
  • Equipped with a tricolor LED interface for status identification.