Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional Advanced Hand Sanitizer Fragrance Free Gel
GREENER_CHOICESDP

Catalog No. 19043803 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
PURELL ES8 Touch-Free Hand Sanitizer Dispensers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-043-803 PURELL ES8 Touch-Free Hand Sanitizer Dispensers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-803 Supplier Gojo Industries Inc Supplier No. 776002
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Purell™ hand sanitizer dispensers.

  • Green certified
  • Fragrance-free gel hand sanitizer for professional environments
  • Kills more than 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce for ounce
  • Made with naturally renewable ethanol
  • Clinically proven formulation to maintain skin health
  • Dye and fragrance free
  • Supports LEED certification criteria
  • AT-A-GLANCE™ refills - Monitor product level with just one look
  • SANITARY SEALED™ PET refill is easily recycled
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sterilizer/Disinfectant
Color Clear
Form Gel
For Use With (Equipment) PURELL ES8 Touch-Free Hand Sanitizer Dispensers
Quantity 2/Pk.
Size 1200 mL
Volume (English) 40.58 oz.
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.