Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Professional Fresh Scent Foam Soap
GREENER_CHOICESDP

Catalog No. 19044054 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-054 1500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-054 Supplier PURELL™ Supplier No. 856104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Professional Fresh Scent Foam Soap

  • Professional
  • Fresh Scent
  • Foam Soap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Soap
Color Clear
Form Foam
Size 1.5 L
Volume (Metric) 1500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.