Learn More
Anton Paar PROFESSIONAL SOFTWARE FOR DSC
Catalog No. 5000580495
Encompass_Preferred
Julia Suite Professional Software package with full analysis potential.All important evaluation features for everyday work are included:• Peak• Glass Transition• Specific Heat Capacity• Oxidation Induction Time• Quality control including control charts• Calibration for sinusoidal DSC incl. SDSC measurements• Measurement/Analysis report as pdfLicense for 1 measurement and 5 analysis installations included
Catalog No. 50-005-80495 Supplier Anton Paar Supplier No. 268008
Julia Suite Professional Software package with full analysis potential.All important evaluation features for everyday work are included:• Peak• Glass Transition• Specific Heat Capacity• Oxidation Induction Time• Quality control including control charts• Calibration for sinusoidal DSC incl. SDSC measurements• Measurement/Analysis report as pdfLicense for 1 measurement and 5 analysis installations included