Learn More
BURKLE INC Profi Sampler Aluminium, complete
Catalog No. 5000025956
Encompass_Preferred
While the MiniSampler is designed for laboratory use, the ProfiSampler has been specially developed for rough industrial use.
Vacuum samplers have the advantage that the sample liquid only comes into contact with the suction hose and the original sample bottle. The possibility of cross-contamination is greatly reduced. Immediately after taking the sample, the bottles are sealed. The disposable suction hose can quickly be replaced. This guarantees that the samples are always authentic. If necessary, a new hose can be used for every single sample.
Due to its small diameter (8 mm (0.31 in.)), the hose can reach narrow, seemingly inaccessible sampling areas.
Catalog No. 50-000-25956 Supplier BURKLE INC Supplier No. 53051100
While the MiniSampler is designed for laboratory use, the ProfiSampler has been specially developed for rough industrial use.
Vacuum samplers have the advantage that the sample liquid only comes into contact with the suction hose and the original sample bottle. The possibility of cross-contamination is greatly reduced. Immediately after taking the sample, the bottles are sealed. The disposable suction hose can quickly be replaced. This guarantees that the samples are always authentic. If necessary, a new hose can be used for every single sample.
Due to its small diameter (8 mm (0.31 in.)), the hose can reach narrow, seemingly inaccessible sampling areas.