Learn More
Hach Company PROFIBUS DP Comm Mod SC200
Supplier:  Hach Company 9173900
Encompass
The Profibus DP communication module for the SC200 is a user installed module that allows for communication via Profibus from the controller See manual for additional connection information
Catalog No. 50-002-07442