Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ PROFILE-II™ Cassette Drug Testing Device for AMP/BAR/BZO/COC/OPI2/PCP/THC

Catalog No. 21500109
Change view
Click to view available options
Quantity:
25 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-500-109 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-500-109 Supplier MEDTOX™ Supplier No. 601607
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Pipette

Conduct on-site screening for up to 11 drugs of abuse including Oxycodone. MEDTOX™ Diagnostics PROFILE-II™ Cassette Drug Testing Devices are easy to use and cost-effective. These visual read, single-assay devices deliver accurate screening results in minutes.

  • 510(k)-cleared
  • Made in the USA
  • Quality management system conforms with ISO 13485:2003
  • Accurate screening results in minutes
  • On-board adulteration strip
  • Panel options that can test for up to 11 drugs – including Oxycodone and Methamphetamine/Ecstasy
  • Technical support services available for all MEDTOX Diagnostics, Inc. products
Screening Thresholds
Amphetamine (AM/AMP): 1000ng/mL

Barbiturates (BA/BAR): 200ng/mL

Benzodiazepine(BZ/BZO): 300ng/mL

Cocaine (CO/COC): 300ng/mL

Methamphetamine (MA/MAMP): 1000ng/mL

Methadone (MTD): 300ng/mL

Opiates (OP/OPI): 2000ng/mL

Oxycodone (OX/OXY): 100ng/mL

Phencyclidine (PC/PCP): 25ng/mL

Propoxyphene (PP/PPX): 300ng/mL

Cannabinoids (TH/THC): 50ng/mL

Specifications

Sample Type Urine
For Use With (Application) Seven panel drugs of abuse test for AMP/BAR/BZO/COC/OPI2/PCP/THC - specimen type is urine
Quantity 25 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.