Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ PROFILE-II ER™ Cassette Drug Testing Device for AMP, BAR, BZO, COC, MTD, OPI, PCP, TCA, THC

Catalog No. 21500104
Change view
Click to view available options
Quantity:
25 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-500-104 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-500-104 Supplier MEDTOX™ Supplier No. 601591
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Pipette

Conduct on-site screening for up to 12 drugs of abuse including Cannabinoids, Cocaine, Oxycodone, Methamphetamine and Opiates. MEDTOX™ Diagnostics PROFILE-II ER™ Cassette Drug Testing Devices are easy to use and cost-effective.

  • Visual read, single-assay devices deliver accurate screening results in minutes
  • 510(k)-cleared
  • Made in the USA
  • Quality management system conforms with ISO 13485:2003
  • Accurate screening results in minutes
  • On-board adulteration strip available on some products
  • Tests for up to 12 drugs of abuse in a single device
  • Technical support services available for all MEDTOX Diagnostics, Inc. products
PROFILE-II ER Screening Thresholds
Amphetamine (AMP): 1000ng/mL

Barbiturates (BAR): 200ng/mL

Benzodiazepine(BZO): 300ng/mL

Cocaine (COC): 300ng/mL

3,4-Methylenedioxy-N-Methylamphetamine (MDMA) 1500ng/mL

Methamphetamine (MAMP): 1000ng/mL

Methadone (MTD): 300ng/mL

Opiates (OPI3): 300ng/mL

Oxycodone (OXY): 100ng/mL

Phencyclidine (PCP): 25ng/mL

Tricyclic Antidepressant (TCA): 300ng/mL

Propoxyphene (PPX): 300ng/mL

Cannabinoids (THC): 50ng/mL

Specifications

Sample Type Urine
For Use With (Application) Nine panel drugs of abuse test for AMP/BAR/BZO/COC/MTD/OPI3/PCP/TCA/THC - specimen type is urine
Quantity 25 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.