Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ Diagnostics PROFILE™-IIA 9 Panel

Catalog No. 16800566
Change view
Click to view available options
Detectable Analytes:
Oxazepam, Benzodiazepines (BZO)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
16-800-566 Oxazepam, Benzodiazepines (BZO)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-800-566 Supplier MEDTOX™ Diagnostics Supplier No. 601683
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PROFILE™-IIA 9 Panel detects AMP/BAR/BZO/COC/MAMP-MDMA/OPI2/OXY/PCP/THC + LFAS.

  • Detects Amphetamine, Barbiturates, Benzodiazepines, Cocaine, Methampehtamine-Methylenedioxymethampheatmine (MDMA), Opiates (2000), Oxycodone, Phencyclidein (PCP), Cannabinoides (THC) + Lateral Flow Adulteration Strip (LFAS).

Specifications

CE Marker No
Clia Complexity Moderate
Detectable Analytes Oxazepam, Benzodiazepines (BZO)
Format Cassette
Sample Type Urine
Sensitivity 500 ng/mL (AMP), 200 ng/mL (BAR), 150 ng/mL (BZO), 10 ng/mL (BUP), 150 ng/mL (COC), 500 ng/mL (MAMP), 200 ng/mL (MTD), 100 ng/mL (OPI), 2000 ng/mL (OPI2), 100 ng/mL (OXY), 25 ng/mL (PCP), 300 ng/mL (PPX), 50 ng/mL (THC), 300 ng/mL (TCA)
Type Drug Test Panel
Disposable Single-use
For Use With (Application) Drug Panel (Urine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.