Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ Diagnostics PROFILE™-V 12 Panel

Catalog No. 21124423
Change view
Click to view available options
Detectable Analytes:
Oxazepam, Benzodiazepines (BZO), Tricyclic Antidepressants (TCA)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
21-124-423 Oxazepam, Benzodiazepines (BZO), Tricyclic Antidepressants (TCA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-124-423 Supplier MEDTOX™ Diagnostics Supplier No. 604032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PROFILE™-V 12 Panel detects AMP/BAR/BUP/BZO/COC/MAMP/MTD/OPI/OXY/PCP/TCA/THC.

  • Detects Amphetamine, Barbiturates, Benzodiazepines, Buprenorphine, Cocaine, Methamphetamine, Methadone, Opiates (100 ng/mL), Oxycodone, Phencyclidine (PCP) , Tricyclic Antidepressants, Cannabinoides (THC).

Specifications

CE Marker No
Clia Complexity Moderate
Detectable Analytes Oxazepam, Benzodiazepines (BZO), Tricyclic Antidepressants (TCA)
Format Cassette
Sample Type Urine
Sensitivity 500 ng/mL (AMP), 200 ng/mL (BAR), 150 ng/mL (BZO), 10 ng/mL (BUP), 150 ng/mL (COC), 500 ng/mL (MAMP), 200 ng/mL (MTD), 100 ng/mL (OPI), 2000 ng/mL (OPI2), 100 ng/mL (OXY), 25 ng/mL (PCP), 300 ng/mL (PPX), 50 ng/mL (THC), 300 ng/mL (TCA)
Type Drug Test Panel
Disposable Single-use
For Use With (Application) Drug Panel (Urine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.