Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ PROFILE™-V DeTipper Bottle

Catalog No. 21500133
Change view
Click to view available options
For Use With (Equipment):
MEDTOX Scan Reader
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-500-133 MEDTOX Scan Reader
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-500-133 Supplier MEDTOX™ Supplier No. 102058
Add to Cart
Add to Cart
This item is not returnable. View return policy

MEDTOX Diagnostics, Inc.™ PROFILE™-V DeTipper Bottle is used to discard used pipette tips from the pipette that is used to dispense urine on to PROFILE-V products.

  • Bottle that stores used disposable pipette tips
  • Order as bottle or each

Specifications

Content And Storage Ambient
For Use With (Equipment) MEDTOX Scan Reader
Type DeTipper Bottle for Used Pipette Tips
Intended Use Used to discard used pipette tips from the pipette that is used to dispense urine on to PROFILE-V products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.