Learn More
Selleck Chemical LLC Proflavine Hemisulfate S4236-50mg
SDP
Catalog No. 503109391
Encompass_Preferred
Proflavine Hemisulfate is a topical antiseptic by interchelating DNA thereby disrupting DNA synthesis and leading to high levels of mutation in the copied DNA strands
Catalog No. 50-310-9391 Supplier Selleck Chemical LLC Supplier No. S423650MG
Proflavine Hemisulfate is a topical antiseptic by interchelating DNA thereby disrupting DNA synthesis and leading to high levels of mutation in the copied DNA strands