Learn More
Apexbio Technology LLC Proflavine Hemisulfate 1811-28-5 50mg
SDP
Catalog No. 5000089931
Encompass_Preferred
Proflavine Hemisulfate (CAS 1811-28-5) is a small-molecule inhibitor targeting DNA It is designed to intercalate into DNA strands thereby disrupting DNA replication and transcription Proflavine Hemisulfate exerts its biological activity primarily through DNA intercalation In in vitro studies Proflavine Hemisulfate demonstrates antimicrobial activity with reported IC50 values for inhibiting bacterial growth typically ranging between 1 10 M depending on the microbial strain and assay conditions used Based on these pharmacological properties Proflavine Hemisulfate holds research potential in studies of antimicrobial activity DNA replication assays mutagenesis and evaluating sensitivity and resistance profiles of microorganisms
Catalog No. 50-000-89931 Supplier Apexbio Technology LLC Supplier No. B182150
Proflavine Hemisulfate (CAS 1811-28-5) is a small-molecule inhibitor targeting DNA It is designed to intercalate into DNA strands thereby disrupting DNA replication and transcription Proflavine Hemisulfate exerts its biological activity primarily through DNA intercalation In in vitro studies Proflavine Hemisulfate demonstrates antimicrobial activity with reported IC50 values for inhibiting bacterial growth typically ranging between 1 10 M depending on the microbial strain and assay conditions used Based on these pharmacological properties Proflavine Hemisulfate holds research potential in studies of antimicrobial activity DNA replication assays mutagenesis and evaluating sensitivity and resistance profiles of microorganisms