Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proflavine hydrochloride, MP Biomedicals™

Catalog No. ICN10360010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10360010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10360010 Supplier MP Biomedicals, Inc Supplier No. 0210360010
Add to Cart
Add to Cart

Chemical Identifiers

CAS 952-23-8
Molecular Formula C26H26Cl2N6O
Molecular Weight (g/mol) 509.44
InChI Key APOVFGFHTIKDOX-UHFFFAOYSA-N
Synonym acridine-3,6-diamine hydrochloride, proflavine hydrochloride, 3,6-diaminoacridine hydrochloride, proflavine monohydrochloride, proflavin hydrochloride, proflavine hcl, 3,6-diaminoacridine hcl, 1,7-diaminoacridine hydrochloride, 3,6-acridinediamine, monohydrochloride, 3,6-diaminoacridine monohydrochloride
PubChem CID 197873
ChEBI CHEBI:74718
IUPAC Name dihydrogen bis(acridine-3,6-diamine) hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1

Specifications

Color Red to Orange
Quantity 10 g
Formula Weight 245.7
Physical Form Powder
Chemical Name or Material Proflavine hydrochloride
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.