Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proflavine Hydrochloride, Spectrum™ Chemical
SDP

Catalog No. 18605840 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-840 1 kg Amber Glass Bottle
18-605-839 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-840 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14281KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1428, 952-23-8, C13H11N3·HCl

Spectrum™ Chemical Proflavine Hydrochloride

Chemical Identifiers

CAS 952-23-8
Molecular Formula C26H26Cl2N6O
Molecular Weight (g/mol) 509.44
InChI Key APOVFGFHTIKDOX-UHFFFAOYSA-N
IUPAC Name dihydrogen bis(acridine-3,6-diamine) hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 kg
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Proflavine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.