Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 1505 Low-Profile Weight Lifters Back Support Brace

Catalog No. 19819466 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-466 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-466 Supplier Ergodyne™ Supplier No. 11494

Low-Profile Weight Lifters Back Support with flexible soft foam core provides support when lifting.

  • 6.25 in. low-profile tapered design
  • Flexible soft foam core composition
  • Nylon jersey body fabric
  • Easy cam buckle closure system
  • Rubber track webbing

Specifications

Color Black
Size X-Large
For Use With (Application) Material Handling, Construction, Warehousing/Distribution, Maintenance
Width (English) 6.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.