Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 2000SF High-Performance Spandex Back Support Brace

Catalog No. 19819457 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
X-Small
2X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-457 3X-Large
19-819-455 X-Small
19-819-456 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-819-457 Supplier Ergodyne™ Supplier No. 11287
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ProFlex™ 2000SF High-Performance Spandex Back Support is designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.

  • Non-conductive Sticky Fingers™ stays for long wear
  • Innovative Power Panel Positioners™ ensure the tension bands stay close by
  • Unique V-shaped design
  • 840D spandex body material
  • Adjustable, detachable suspenders keep back brace in place
  • Waist size ranges from 25 to 58 in. (64 to 147 cm)
  • Machine wash cold, air dry

Specifications

Color Black
Size 3X-Large
For Use With (Application) Material Handling, Construction, Warehousing/Distribution, Maintenance
Material 840D Spandex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.