Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 355 Short Cap Injected Gel Knee Pads

Catalog No. 19820288 Shop All Ergodyne Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-288 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-288 Supplier Ergodyne™ Supplier No. 18455

ProFlex™ 355 Short Cap Injected Gel Knee Pads provide injected gel comfort and durability.

  • Short cap design for mobility when kneeling
  • Non-slip rubber cap grips kneeling surface
  • Injected gel and PU foam for all-day comfort
  • Bottom webbing and top elastic straps
  • Plastic clips cinch tight and keep straps secure

Specifications

Color Black
Design Short Cap
For Use With (Application) Construction, Maintenance, Mechanics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.