Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 377 Hard Cap Lightweight Gel Knee Pads

Catalog No. 19820292 Shop All Ergodyne Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-292 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-292 Supplier Ergodyne™ Supplier No. 18477

ProFlex™ 377 Hard Cap Lightweight Gel Knee Pads allow you to work longer without overstressing your joints.

  • Abrasion-resistant round hard-cap for long-lasting durability and versatility
  • Smooth hard-cap makes it easy to slide over various surfaces
  • Lightweight gel and EVA foam provide mobile, long-lasting comfort and stability 
  • Breathable perforated neoprene bottom strap provides easy adjustment and secure fit
  • Easy adjustable top elastic strap cinches tight with plastic clip
  • One size fits most

Specifications

Color Black
Design Round, Hard Cap
For Use With (Application) Landscapers, Gardeners, Plumbers, Electricians
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.